Assay Details
| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027168.2 | NP_081444.1 | 1 - 2 | 81 | 83 | |
| GenBank mRNA | BC104360.1 | - | 1 - 2 | 81 | 40049707 | 83 |
| BC104361.1 | - | 1 - 2 | 81 | 40049709 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hddc2
Genomic Map