Assay Details
| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027168.2 | NP_081444.1 | 4 - 5 | 373 | 77 | |
| GenBank mRNA | AK009957.1 | - | 4 - 5 | 314 | 77 | |
| BC104360.1 | - | 4 - 5 | 373 | 40049707 | 77 | |
| BC104361.1 | - | 4 - 5 | 373 | 40049709 | 77 | |
More Information
Set Membership: |
|

Hddc2
Genomic Map