Assay Details
| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027168.2 | NP_081444.1 | 5 - 6 | 513 | 68 | |
| GenBank mRNA | AK009957.1 | - | 5 - 6 | 454 | 68 | |
| BC104360.1 | - | 5 - 6 | 513 | 40049707 | 68 | |
| BC104361.1 | - | 5 - 6 | 513 | 40049709 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27043768 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27043770 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004930 | NM_027168.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004930 | AK009957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004930 | BC104360.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004930 | BC104361.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779621 | AK008052.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779621 | AK136525.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779621 | AK153247.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779621 | BC003493.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779621 | NM_053217.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 768 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76774 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160544 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117927 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004930 | NM_027168.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004930 | AK009957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004930 | BC104360.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004930 | BC104361.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27043781 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117927 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160544 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07019021 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02096251 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160544 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117927 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27043768 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27043770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27043810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08517742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08231087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08808986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08259956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08909110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08259961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08428784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08696423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08727053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08808989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08712333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08712336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27043791 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10362399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362400 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428823_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10362394 | - | - |
| Mouse Genome 430 2.0 Array | 1428823_at | - | - |
| Mouse Genome 430 2.0 Array | 1428823_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69692_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69692_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000001874.mm.1 | NM_027168.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001874.mm.1 | NM_027168.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001874.mm.1 | TC1000000279.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001875.mm.1 | NM_027168.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001875.mm.1 | NM_027168.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001875.mm.1 | TC1000000279.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428823_at | - | - |
| Mouse Expression Set 430 | 1428823_PM_at | - | - |
| Murine Genome U74v2 Set | 160659_PM_at | - | - |
| Murine Genome U74v2 Set | 160659_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428823_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428823_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428823_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428823_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 830184 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244187 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2638781 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244187 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2638781 | - | - |

Hddc2
Genomic Map