Assay Details
| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029912.2 | NP_001025083.1 | 4 - 5 | 1314 | 132 | |
| GenBank mRNA | AK122519.1 | - | 4 - 5 | 1314 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zswim5
Genomic Map