Assay Details
| Gene Symbol: Nlrp5-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045120.1 | 12 - 13 | 1549 | 121 | ||
| GenBank mRNA | AK016782.1 | - | 12 - 13 | 1550 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp5-ps
Genomic Map