Assay Details
| Gene Symbol: Szt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198170.4 | NP_937813.3 | 40 - 41 | 5877 | 59 | |
| GenBank mRNA | AK129152.1 | - | 4 - 5 | 470 | 59 | |
| FJ998170.1 | - | 40 - 41 | 5877 | 59 | ||
| GU433214.1 | - | 40 - 41 | 5803 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Szt2
Genomic Map