Assay Details
| Gene Symbol: Asap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008232.2 | NP_001008233.1 | 5 - 6 | 628 | 59 | |
| GenBank mRNA | AK132294.1 | - | 5 - 6 | 597 | 59 | |
| BC085250.1 | - | 5 - 6 | 581 | 30730459 | 59 | |
More Information
Set Membership: |
|

Asap3
Genomic Map