Assay Details
| Gene Symbol: H2afv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029938.1 | NP_084214.1 | 2 - 3 | 181 | 77 | |
| GenBank mRNA | AK021267.1 | - | 2 - 3 | 104 | 77 | |
| AK155105.1 | - | 2 - 3 | 184 | 77 | ||
| BC147293.1 | - | 1 - 2 | 140 | 9056353 | 77 | |
| BC147294.1 | - | 2 - 3 | 166 | 9056344 | 77 | |
| BC147741.1 | - | 2 - 3 | 166 | 9056690 | 77 | |
| BC147751.1 | - | 2 - 3 | 166 | 9056700 | 77 | |
| CJ069840.1 | - | 2 - 3 | 182 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H2afv
Genomic Map