Assay Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034858.3 | NP_001030030.2 | 1 - 2 | 190 | 104 | |
| GenBank mRNA | AK011447.1 | - | 1 - 2 | 189 | 104 | |
| BM942469.1 | - | 1 - 2 | 173 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011447.1, NM_001034858.3 transcripts.
Set Membership: |
|

Armc2
Genomic Map