Assay Details
| Gene Symbol: Mmrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163507.1 | NP_001156979.1 | 2 - 3 | 941 | 95 | |
| NM_027613.1 | NP_081889.1 | 2 - 3 | 941 | 95 | ||
| GenBank mRNA | AK036724.1 | - | 2 - 3 | 779 | 95 | |
| AK137556.1 | - | 2 - 3 | 780 | 95 | ||
| BC137623.1 | - | 2 - 3 | 741 | 8860642 | 95 | |
| BC144789.1 | - | 2 - 3 | 741 | 9053337 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmrn1
Genomic Map