Assay Details
| Gene Symbol: Mmrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027613.1 | NP_081889.1 | 6 - 7 | 3296 | 66 | |
| GenBank mRNA | AJ586616.1 | - | 1 - 2 | 266 | 66 | |
| AK036724.1 | - | 6 - 7 | 3135 | 66 | ||
| BC046425.1 | - | 1 - 2 | 596 | 66 | ||
| BC137623.1 | - | 6 - 7 | 3096 | 8860642 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmrn1
Genomic Map