Assay Details
| Gene Symbol: Ccser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183310.2 | NP_899133.2 | 1 - 2 | 330 | 101 | |
| GenBank mRNA | BB634602.1 | - | 1 - 2 | 291 | 101 | |
| BC107398.2 | - | 1 - 2 | 117 | 40058350 | 101 | |
| BC107399.1 | - | 1 - 2 | 116 | 40058352 | 101 | |
| BC117738.2 | - | 1 - 2 | 117 | 40089965 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107398.2, BC107399.1, BC117738.2, NM_183310.2 transcripts.
Set Membership: |
|

Ccser1
Genomic Map