| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600005579.mm.1 |
TC0600000725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005579.mm.1 |
NM_001164316.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005579.mm.1 |
NM_001164316.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005579.mm.1 |
NM_183310.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005579.mm.1 |
NM_183310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005578.mm.1 |
TC0600000725.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005578.mm.1 |
NM_001164316.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005578.mm.1 |
NM_001164316.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005578.mm.1 |
NM_183310.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005578.mm.1 |
NM_183310.2 |
Refseq |