Assay Details
| Gene Symbol: Ccser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164316.1 | NP_001157788.1 | 4 - 5 | 1914 | 107 | |
| NM_183310.2 | NP_899133.2 | 4 - 5 | 1966 | 107 | ||
| GenBank mRNA | AK173231.1 | - | 3 - 4 | 659 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccser1
Genomic Map