Assay Details
| Gene Symbol: Cyp4a30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100185.1 | NP_001093655.1 | 6 - 7 | 787 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4a30b| Gene Symbol: Cyp4a30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100185.1 | NP_001093655.1 | 6 - 7 | 787 | 67 | |
Set Membership: |
|