| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400029421.mm.1 |
TC0400003605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029421.mm.1 |
NM_001195730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029421.mm.1 |
NM_001195730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029423.mm.1 |
TC0400003605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029423.mm.1 |
NM_001195730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029423.mm.1 |
NM_001195730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029422.mm.1 |
TC0400003605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029422.mm.1 |
NM_001195730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029422.mm.1 |
NM_001195730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029420.mm.1 |
TC0400003605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029420.mm.1 |
NM_001195730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029420.mm.1 |
NM_001195730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029417.mm.1 |
TC0400003605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029417.mm.1 |
NM_001195730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029417.mm.1 |
NM_001195730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029419.mm.1 |
TC0400003605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029419.mm.1 |
NM_001195730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029419.mm.1 |
NM_001195730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029426.mm.1 |
TC0400003605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029426.mm.1 |
NM_001195730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029426.mm.1 |
NM_001195730.1 |
Refseq |