Assay Details
| Gene Symbol: Abcg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011920.3 | NP_036050.1 | 14 - 15 | 2193 | 99 | |
| GenBank mRNA | AF103875.1 | - | 2 - 3 | 207 | 99 | |
| AF140218.1 | - | 13 - 14 | 1756 | 99 | ||
| BC053730.1 | - | 14 - 15 | 2205 | 30064535 | 99 | |
| KF662864.1 | - | 14 - 15 | 1790 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcg2
Genomic Map