Assay Details
| Gene Symbol: Smarcad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253392.1 | NP_001240321.1 | 14 - 15 | 1951 | 86 | |
| NM_007958.1 | NP_031984.1 | 16 - 17 | 2412 | 86 | ||
| GenBank mRNA | AK122454.1 | - | 16 - 17 | 2219 | 86 | |
| AK134442.1 | - | 16 - 17 | 2263 | 86 | ||
| BC042442.1 | - | 14 - 15 | 1944 | 3980726 | 86 | |
| X69942.1 | - | 16 - 17 | 2412 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smarcad1
Genomic Map