Assay Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027547.2 | NP_081823.2 | 2 - 3 | 318 | 105 | |
| GenBank mRNA | AK014501.1 | - | 2 - 3 | 242 | 105 | |
| AK018310.1 | - | 2 - 3 | 256 | 105 | ||
| BC089598.1 | - | 2 - 3 | 258 | 105 | ||
| BC138901.1 | - | 2 - 3 | 292 | 8861923 | 105 | |
| BC138902.1 | - | 2 - 3 | 292 | 8861924 | 105 | |
| BY014014.1 | - | 2 - 3 | 319 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdm5
Genomic Map