Assay Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168304.1 | NP_001161776.1 | 11 - 12 | 1399 | 93 | |
| NM_001291816.1 | NP_001278745.1 | 10 - 11 | 1323 | 93 | ||
| NM_001291817.1 | NP_001278746.1 | 10 - 11 | 1341 | 93 | ||
| NM_198164.3 | NP_937807.2 | 10 - 11 | 1267 | 93 | ||
| GenBank mRNA | AK028643.1 | - | 968 | 93 | ||
| AK052818.1 | - | 11 - 12 | 1230 | 93 | ||
| AK053651.1 | - | 10 - 11 | 1240 | 93 | ||
| AK122421.1 | - | 11 - 12 | 1399 | 93 | ||
| AK147340.1 | - | 10 - 11 | 1121 | 93 | ||
| BC030312.1 | - | 4 - 5 | 295 | 93 | ||
| BC062100.1 | - | 10 - 11 | 1123 | 93 | ||
| BC080280.1 | - | 10 - 11 | 1273 | 93 | ||
| BC137614.1 | - | 11 - 12 | 1353 | 8860633 | 93 | |
| BC144782.1 | - | 11 - 12 | 1353 | 9053330 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk19
Genomic Map