Assay Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168304.1 | NP_001161776.1 | 2 - 3 | 488 | 70 | |
| NM_001291817.1 | NP_001278746.1 | 2 - 3 | 488 | 70 | ||
| NM_198164.3 | NP_937807.2 | 2 - 3 | 488 | 70 | ||
| GenBank mRNA | AK028643.1 | - | 115 | 70 | ||
| AK052818.1 | - | 2 - 3 | 319 | 70 | ||
| AK053651.1 | - | 2 - 3 | 461 | 70 | ||
| AK122421.1 | - | 2 - 3 | 488 | 70 | ||
| BC048908.1 | - | 2 - 3 | 238 | 70 | ||
| BC062100.1 | - | 2 - 3 | 270 | 70 | ||
| BC137614.1 | - | 2 - 3 | 442 | 8860633 | 70 | |
| BC144782.1 | - | 2 - 3 | 442 | 9053330 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291817.1 transcripts.
Set Membership: |
|

Cdk19
Genomic Map