Assay Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168304.1 | NP_001161776.1 | 3 - 4 | 602 | 68 | |
| NM_001291816.1 | NP_001278745.1 | 2 - 3 | 526 | 68 | ||
| NM_001291817.1 | NP_001278746.1 | 3 - 4 | 602 | 68 | ||
| NM_198164.3 | NP_937807.2 | 3 - 4 | 602 | 68 | ||
| GenBank mRNA | AK028643.1 | - | 229 | 68 | ||
| AK052818.1 | - | 3 - 4 | 433 | 68 | ||
| AK053651.1 | - | 3 - 4 | 575 | 68 | ||
| AK122421.1 | - | 3 - 4 | 602 | 68 | ||
| AK147340.1 | - | 2 - 3 | 324 | 68 | ||
| BC048908.1 | - | 3 - 4 | 352 | 68 | ||
| BC062100.1 | - | 3 - 4 | 384 | 68 | ||
| BC080280.1 | - | 2 - 3 | 476 | 68 | ||
| BC137614.1 | - | 3 - 4 | 556 | 8860633 | 68 | |
| BC144782.1 | - | 3 - 4 | 556 | 9053330 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291816.1, NM_001291817.1 transcripts.
Set Membership: |
|

Cdk19
Genomic Map