Assay Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168304.1 | NP_001161776.1 | 4 - 5 | 739 | 78 | |
| NM_001291816.1 | NP_001278745.1 | 3 - 4 | 663 | 78 | ||
| NM_198164.3 | NP_937807.2 | 4 - 5 | 739 | 78 | ||
| GenBank mRNA | AK052818.1 | - | 4 - 5 | 570 | 78 | |
| AK053651.1 | - | 4 - 5 | 712 | 78 | ||
| AK122421.1 | - | 4 - 5 | 739 | 78 | ||
| AK147340.1 | - | 3 - 4 | 461 | 78 | ||
| BC048908.1 | - | 4 - 5 | 489 | 78 | ||
| BC080280.1 | - | 3 - 4 | 613 | 78 | ||
| BC137614.1 | - | 4 - 5 | 693 | 8860633 | 78 | |
| BC144782.1 | - | 4 - 5 | 693 | 9053330 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk19
Genomic Map