Assay Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168304.1 | NP_001161776.1 | 7 - 8 | 1080 | 109 | |
| NM_001291816.1 | NP_001278745.1 | 6 - 7 | 1004 | 109 | ||
| NM_001291817.1 | NP_001278746.1 | 6 - 7 | 1022 | 109 | ||
| NM_198164.3 | NP_937807.2 | 6 - 7 | 948 | 109 | ||
| GenBank mRNA | AK028643.1 | - | 649 | 109 | ||
| AK052818.1 | - | 7 - 8 | 911 | 109 | ||
| AK122421.1 | - | 7 - 8 | 1080 | 109 | ||
| AK147340.1 | - | 6 - 7 | 802 | 109 | ||
| BC048908.1 | - | 7 - 8 | 830 | 109 | ||
| BC062100.1 | - | 6 - 7 | 804 | 109 | ||
| BC080280.1 | - | 6 - 7 | 954 | 109 | ||
| BC137614.1 | - | 7 - 8 | 1034 | 8860633 | 109 | |
| BC144782.1 | - | 7 - 8 | 1034 | 9053330 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk19
Genomic Map