Assay Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168304.1 | NP_001161776.1 | 8 - 9 | 1145 | 85 | |
| NM_001291816.1 | NP_001278745.1 | 7 - 8 | 1069 | 85 | ||
| NM_001291817.1 | NP_001278746.1 | 7 - 8 | 1087 | 85 | ||
| NM_198164.3 | NP_937807.2 | 7 - 8 | 1013 | 85 | ||
| GenBank mRNA | AK028643.1 | - | 714 | 85 | ||
| AK052818.1 | - | 8 - 9 | 976 | 85 | ||
| AK053651.1 | - | 7 - 8 | 986 | 85 | ||
| AK122421.1 | - | 8 - 9 | 1145 | 85 | ||
| AK147340.1 | - | 7 - 8 | 867 | 85 | ||
| BC048908.1 | - | 8 - 9 | 895 | 85 | ||
| BC062100.1 | - | 7 - 8 | 869 | 85 | ||
| BC080280.1 | - | 7 - 8 | 1019 | 85 | ||
| BC137614.1 | - | 8 - 9 | 1099 | 8860633 | 85 | |
| BC144782.1 | - | 8 - 9 | 1099 | 9053330 | 85 | |
More Information
Set Membership: |
|

Cdk19
Genomic Map