Assay Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168304.1 | NP_001161776.1 | 9 - 10 | 1223 | 67 | |
| NM_001291816.1 | NP_001278745.1 | 8 - 9 | 1147 | 67 | ||
| NM_001291817.1 | NP_001278746.1 | 8 - 9 | 1165 | 67 | ||
| NM_198164.3 | NP_937807.2 | 8 - 9 | 1091 | 67 | ||
| GenBank mRNA | AK028643.1 | - | 792 | 67 | ||
| AK052818.1 | - | 9 - 10 | 1054 | 67 | ||
| AK053651.1 | - | 8 - 9 | 1064 | 67 | ||
| AK122421.1 | - | 9 - 10 | 1223 | 67 | ||
| AK147340.1 | - | 8 - 9 | 945 | 67 | ||
| BC030312.1 | - | 2 - 3 | 119 | 67 | ||
| BC048908.1 | - | 9 - 10 | 973 | 67 | ||
| BC062100.1 | - | 8 - 9 | 947 | 67 | ||
| BC080280.1 | - | 8 - 9 | 1097 | 67 | ||
| BC137614.1 | - | 9 - 10 | 1177 | 8860633 | 67 | |
| BC144782.1 | - | 9 - 10 | 1177 | 9053330 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk19
Genomic Map