Assay Details
| Gene Symbol: Hk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013820.3 | NP_038848.1 | 6 - 7 | 1168 | 67 | |
| GenBank mRNA | AJ238540.1 | - | 6 - 7 | 1111 | 67 | |
| AK145936.1 | - | 6 - 7 | 1149 | 67 | ||
| BC054472.1 | - | 6 - 7 | 1087 | 6465367 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hk2
Genomic Map