Assay Details
| Gene Symbol: Sim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011376.3 | NP_035506.2 | 9 - 10 | 1407 | 93 | |
| GenBank mRNA | AB221630.1 | - | 9 - 10 | 1161 | 93 | |
| D79209.1 | - | 9 - 10 | 1200 | 93 | ||
| U40575.1 | - | 9 - 10 | 1404 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sim1
Genomic Map