Assay Details
| Gene Symbol: Mast2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042743.2 | NP_001036208.1 | 2 - 3 | 591 | 67 | |
| NM_008641.3 | NP_032667.2 | 2 - 3 | 591 | 67 | ||
| GenBank mRNA | BC060703.1 | - | 2 - 3 | 161 | 6412811 | 67 |
| CB246117.1 | - | 2 - 3 | 332 | 67 | ||
| U02313.1 | - | 2 - 3 | 258 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mast2
Genomic Map