Assay Details
| Gene Symbol: Txnrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178058.1 | NP_001171529.1 | 13 - 14 | 1805 | 79 | |
| NM_001178059.1 | NP_001171530.1 | 11 - 12 | 1463 | 79 | ||
| NM_001178060.1 | NP_001171531.1 | 11 - 12 | 1463 | 79 | ||
| NM_153162.3 | NP_694802.2 | 13 - 14 | 1805 | 79 | ||
| GenBank mRNA | AF349659.1 | - | 13 - 14 | 1714 | 79 | |
| AK012699.1 | - | 13 - 14 | 1798 | 79 | ||
| AK041046.1 | - | 13 - 14 | 1808 | 79 | ||
| AK080362.1 | - | 13 - 14 | 1796 | 79 | ||
| AK134987.1 | - | 11 - 12 | 1454 | 79 | ||
| AK200851.1 | - | 2 - 3 | 241 | 79 | ||
| AK218651.1 | - | 2 - 3 | 227 | 79 | ||
| AK220140.1 | - | 2 - 3 | 227 | 79 | ||
| BC076605.1 | - | 13 - 14 | 1763 | 30628949 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd3
Genomic Map