Assay Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033929.2 | NP_001029101.1 | 4 - 5 | 675 | 76 | |
| NM_178413.5 | NP_848500.3 | 5 - 6 | 743 | 76 | ||
| GenBank mRNA | BC025604.1 | - | 5 - 6 | 706 | 5250319 | 76 |
| BC051244.1 | - | 4 - 5 | 637 | 4975661 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Thnsl2
Genomic Map