Assay Details
| Gene Symbol: Cdca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026560.4 | NP_080836.3 | 3 - 4 | 501 | 121 | |
| GenBank mRNA | AK010905.1 | - | 3 - 4 | 493 | 121 | |
| AK020070.1 | - | 3 - 4 | 457 | 121 | ||
| AK076065.1 | - | 3 - 4 | 458 | 121 | ||
| AK076422.1 | - | 3 - 4 | 491 | 121 | ||
| AK077717.1 | - | 3 - 4 | 454 | 121 | ||
| AK077920.1 | - | 3 - 4 | 424 | 121 | ||
| AK078404.1 | - | 3 - 4 | 454 | 121 | ||
| AK082800.1 | - | 3 - 4 | 466 | 121 | ||
| AY550907.1 | - | 3 - 4 | 491 | 121 | ||
| BC068181.1 | - | 3 - 4 | 449 | 30634239 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdca8
Genomic Map