Assay Details
| Gene Symbol: Cdca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026560.4 | NP_080836.3 | 5 - 6 | 688 | 145 | |
| GenBank mRNA | AK076065.1 | - | 5 - 6 | 645 | 145 | |
| AK076422.1 | - | 5 - 6 | 678 | 145 | ||
| AK077717.1 | - | 5 - 6 | 641 | 145 | ||
| AK077920.1 | - | 5 - 6 | 611 | 145 | ||
| AK082800.1 | - | 5 - 6 | 653 | 145 | ||
| AY550907.1 | - | 5 - 6 | 678 | 145 | ||
| BC068181.1 | - | 602 | 30634239 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdca8
Genomic Map