Assay Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008779.2 | NP_032805.2 | 12 - 13 | 1747 | 123 | |
| GenBank mRNA | AK140747.1 | - | 3 - 4 | 336 | 123 | |
| L01991.1 | - | 1856 | 123 | |||
More Information
Set Membership: |
|

Cntn3
Genomic Map