Assay Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008779.2 | NP_032805.2 | 9 - 10 | 1297 | 84 | |
| GenBank mRNA | L01991.1 | - | 1406 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cntn3
Genomic Map