Assay Details
| Gene Symbol: Shq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012978.1 | - | 4 - 5 | 583 | 92 | |
| BC052657.1 | - | 4 - 5 | 575 | 30042589 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shq1
Genomic Map