Assay Details
| Gene Symbol: Cntn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017383.3 | NP_059079.2 | 5 - 6 | 763 | 83 | |
| GenBank mRNA | AB032602.1 | - | 4 - 5 | 449 | 83 | |
| AK052972.1 | - | 6 - 7 | 656 | 83 | ||
| BC076594.1 | - | 5 - 6 | 763 | 30649661 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntn6
Genomic Map