Assay Details
| Gene Symbol: Trh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009426.3 | NP_033452.2 | 1 - 2 | 104 | 57 | |
| GenBank mRNA | AK010666.1 | - | 1 - 2 | 103 | 57 | |
| BC053493.1 | - | 1 - 2 | 90 | 30010395 | 57 | |
| CD562178.1 | - | 1 - 2 | 93 | 57 | ||
| CF951860.1 | - | 1 - 2 | 92 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC053493.1, NM_009426.3 transcripts.
Set Membership: |
|

Trh
Genomic Map