Assay Details
| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011630.3 | NP_035760.1 | 9 - 10 | 1331 | 81 | |
| GenBank mRNA | AK157305.1 | - | 9 - 10 | 1305 | 81 | |
| AK220507.1 | - | 2 - 3 | 229 | 81 | ||
| BC103683.1 | - | 7 - 8 | 950 | 40047943 | 81 | |
| BC103684.1 | - | 7 - 8 | 950 | 40047942 | 81 | |
| BC103686.1 | - | 7 - 8 | 950 | 40047941 | 81 | |
| S75970.1 | - | 9 - 10 | 1446 | 81 | ||
| U11688.1 | - | 8 - 9 | 993 | 81 | ||
| U32939.1 | - | 7 - 8 | 964 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr2c2
Genomic Map