Assay Details
| Gene Symbol: Mitf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113198.1 | NP_001106669.1 | 3 - 4 | 711 | 87 | |
| NM_001178049.1 | NP_001171520.1 | 3 - 4 | 598 | 87 | ||
| NM_008601.3 | NP_032627.1 | 2 - 3 | 402 | 87 | ||
| GenBank mRNA | AB221657.1 | - | 2 - 3 | 264 | 87 | |
| AK052895.1 | - | 3 - 4 | 570 | 87 | ||
| AK154189.1 | - | 3 - 4 | 712 | 87 | ||
| AK155350.1 | - | 3 - 4 | 712 | 87 | ||
| AK200280.1 | - | 1 - 2 | 68 | 87 | ||
| BC108976.2 | - | 2 - 3 | 366 | 40047440 | 87 | |
| Z23066.1 | - | 2 - 3 | 393 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mitf
Genomic Map