Assay Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF327856.1 | - | 1 - 2 | 213 | 125 | |
| AK143978.1 | - | 1 - 2 | 212 | 125 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmd4b| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF327856.1 | - | 1 - 2 | 213 | 125 | |
| AK143978.1 | - | 1 - 2 | 212 | 125 | ||
Set Membership: |
|