Assay Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145148.2 | NP_660130.2 | 11 - 12 | 1221 | 82 | |
| GenBank mRNA | AB042027.1 | - | 11 - 12 | 1144 | 82 | |
| AF327856.1 | - | 10 - 11 | 844 | 82 | ||
| AF327857.1 | - | 9 - 10 | 631 | 82 | ||
| AK129262.1 | - | 10 - 11 | 696 | 82 | ||
| AK132280.1 | - | 11 - 12 | 1085 | 82 | ||
| BC052390.1 | - | 11 - 12 | 1090 | 5718942 | 82 | |
More Information
Set Membership: |
|

Frmd4b
Genomic Map