Assay Details
| Gene Symbol: Sfxn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178639.4 | NP_848754.1 | 9 - 10 | 547 | 69 | |
| GenBank mRNA | AK036285.1 | - | 9 - 10 | 545 | 69 | |
| AK043706.1 | - | 9 - 10 | 546 | 69 | ||
| AK082073.1 | - | 9 - 10 | 545 | 69 | ||
More Information
Set Membership: |
|

Sfxn5
Genomic Map