Assay Details
| Gene Symbol: Rad18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167730.1 | NP_001161202.1 | 7 - 8 | 967 | 72 | |
| NM_021385.2 | NP_067360.2 | 7 - 8 | 967 | 72 | ||
| GenBank mRNA | AF205278.1 | - | 7 - 8 | 891 | 72 | |
| AK012795.1 | - | 7 - 8 | 968 | 72 | ||
| AK028505.1 | - | 7 - 8 | 887 | 72 | ||
| AK030043.1 | - | 7 - 8 | 968 | 72 | ||
| AK048975.1 | - | 7 - 8 | 942 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rad18
Genomic Map