Assay Details
| Gene Symbol: Rassf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178045.4 | NP_835146.3 | 1 - 2 | 118 | 58 | |
| GenBank mRNA | AK036528.1 | - | 1 - 2 | 82 | 58 | |
| AK053578.1 | - | 1 - 2 | 80 | 58 | ||
| AK089460.1 | - | 1 - 2 | 75 | 58 | ||
| AK139284.1 | - | 1 - 2 | 65 | 58 | ||
| AK151561.1 | - | 1 - 2 | 82 | 58 | ||
| AK152639.1 | - | 1 - 2 | 82 | 58 | ||
| AK160584.1 | - | 1 - 2 | 92 | 58 | ||
| AK164357.1 | - | 1 - 2 | 81 | 58 | ||
| AK170955.1 | - | 1 - 2 | 67 | 58 | ||
| AK171198.1 | - | 1 - 2 | 96 | 58 | ||
| BC023245.1 | - | 1 - 2 | 71 | 4459337 | 58 | |
| BC060709.1 | - | 1 - 2 | 73 | 6409692 | 58 | |
| BY170346.1 | - | 1 - 2 | 119 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036528.1, AK139284.1, AK160584.1, AK164357.1, AK170955.1, AK171198.1, BC023245.1, BC060709.1, NM_178045.4 transcripts.
Set Membership: |
|

Rassf4
Genomic Map