Assay Details
| Gene Symbol: Ggcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019802.5 | NP_062776.1 | 5 - 6 | 718 | 70 | |
| GenBank mRNA | AF087938.1 | - | 5 - 6 | 693 | 70 | |
| AK033496.1 | - | 5 - 6 | 645 | 70 | ||
| AK036577.1 | - | 5 - 6 | 692 | 70 | ||
| AK135425.1 | - | 5 - 6 | 705 | 70 | ||
| AK165982.1 | - | 5 - 6 | 693 | 70 | ||
| BC138763.1 | - | 5 - 6 | 643 | 8861785 | 70 | |
| BC138764.1 | - | 5 - 6 | 643 | 8861786 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ggcx
Genomic Map