Assay Details
| Gene Symbol: Vamp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016794.3 | NP_058074.2 | 1 - 2 | 194 | 77 | |
| GenBank mRNA | AF045661.1 | - | 1 - 2 | 69 | 77 | |
| AF053724.1 | - | 1 - 2 | 157 | 77 | ||
| AK002268.1 | - | 1 - 2 | 52 | 77 | ||
| AK013279.1 | - | 1 - 2 | 102 | 77 | ||
| BC012668.1 | - | 1 - 2 | 120 | 4205509 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF045661.1, AF053724.1, AK002268.1, AK013279.1, BC012668.1, NM_016794.3 transcripts.
Set Membership: |
|

Vamp8
Genomic Map