Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600006519.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006519.mm.1 |
NM_007599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006521.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006521.mm.1 |
NM_001271415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006525.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006525.mm.1 |
NM_001042534.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006538.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006538.mm.1 |
NM_001042534.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006538.mm.1 |
NM_001271395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006538.mm.1 |
NM_001271415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006538.mm.1 |
NM_007599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006543.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006543.mm.1 |
NM_001042534.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006543.mm.1 |
NM_001271395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006543.mm.1 |
NM_001271415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006543.mm.1 |
NM_007599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006520.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006520.mm.1 |
NM_007599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006522.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006522.mm.1 |
NM_001271415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006541.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006541.mm.1 |
NM_001042534.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006541.mm.1 |
NM_001271395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006541.mm.1 |
NM_001271415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006541.mm.1 |
NM_007599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006542.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006542.mm.1 |
NM_001042534.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006542.mm.1 |
NM_001271395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006542.mm.1 |
NM_001271415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006542.mm.1 |
NM_007599.4 |
Refseq |