Assay Details
| Gene Symbol: Sh2d6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016886.1 | - | 3 - 4 | 355 | 94 | |
| BC100525.1 | - | 3 - 4 | 312 | 30920200 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC100525.1 transcripts.
Set Membership: |
|

Sh2d6
Genomic Map