Assay Details
| Gene Symbol: Sh2d6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016886.1 | - | 5 - 6 | 429 | 71 | |
| BC100525.1 | - | 5 - 6 | 386 | 30920200 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh2d6
Genomic Map